3-(4-Carboxyphenyl)propionic acid 98%
Dodavatel: Apollo Scientific
Synonyms:
4-(2-Carboxyethyl)benzoic Acid
OR925442-25G
APOSOR925442-25GEA
4800
CZK
APOSOR925442-25G
3-(4-Carboxyphenyl)propionic acid 98%
3-(4-Carboxyphenyl)propionic acid
Vzorec:
C₁₀H₁₀O₄ MW: 194,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016560 CAS číslo: 38628-51-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...