Tisk…

N-Methyl-1-phenylpropylamine 95%

Dodavatel: Apollo Scientific

OR925403-10G OR925403-1G OR925403-5G
APOSOR925403-10GEA 27480 CZK
APOSOR925403-10G APOSOR925403-1G APOSOR925403-5G
N-Methyl-1-phenylpropylamine 95%
N-Methyl-1-phenylpropylamine
Vzorec: C₁₀H₁₅N
Storage Temperature: Ambient
MDL Number: MFCD04038424
CAS číslo: 7713-71-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR