Tisk…

4-Phenylbutyryl chloride 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Phenylbutanoylchloride

OR925349-1G OR925349-5G
APOSOR925349-1GEA 2110 CZK
APOSOR925349-1G APOSOR925349-5G
4-Phenylbutyryl chloride 95%
4-Phenylbutyryl chloride
Vzorec: C₁₀H₁₁ClO
Storage Temperature: Ambient
MDL Number: MFCD00798080
CAS číslo: 18496-54-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR