Tisk…

2-Methylbenzoxazole 95%

Dodavatel: Apollo Scientific

Synonyms: 2-Methyl-1,3-benzoxazole, 2-Methylbenzo[d]oxazole

OR925296-500G
APOSOR925296-500GEA 10680 CZK
APOSOR925296-500G
2-Methylbenzoxazole 95%
2-Methylbenzoxazole
Vzorec: C₈H₇NO
MW: 133,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005771
CAS číslo: 95-21-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR