6-Hydroxy-3-methylpicolinic acid 95%
Dodavatel: Apollo Scientific
OR925285-100MG
OR925285-1G
OR925285-250MG
APOSOR925285-100MGEA
4570
CZK
APOSOR925285-100MG
APOSOR925285-1G
APOSOR925285-250MG
6-Hydroxy-3-methylpicolinic acid 95%
6-Hydroxy-3-methylpicolinic acid
Vzorec:
C₇H₇NO₃ Storage Temperature: Ambient |
CAS číslo:
115185-81-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...