Tisk…

3-Chloro-2,6-xylidine hydrochloride 97%

Dodavatel: Apollo Scientific

OR925208-25G OR925208-5G
APOSOR925208-25GEA 5390 CZK
APOSOR925208-25G APOSOR925208-5G
3-Chloro-2,6-xylidine hydrochloride 97%
3-Chloro-2,6-xylidine hydrochloride
Vzorec: C₈H₁₁Cl₂N
Storage Temperature: Ambient
MDL Number: MFCD02683075
CAS číslo: 72725-98-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR