Tisk…

3,4-Dichloropyridazine

Dodavatel: Apollo Scientific

OR925203-1G OR925203-500MG
APOSOR925203-1GEA 22980 CZK
APOSOR925203-1G APOSOR925203-500MG
3,4-Dichloropyridazine
3,4-Dichloropyridazine
Vzorec: C₄H₂Cl₂N₂
MW: 148,978 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20040517
CAS číslo: 1677-80-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR