Tisk…

3-Methylphthalic anhydride 98%

Dodavatel: Apollo Scientific

Synonyms: 4-Methylisobenzofuran-1,3-dione

OR925180-25G
APOSOR925180-25GEA 13000 CZK
APOSOR925180-25G
3-Methylphthalic anhydride 98%
3-Methylphthalic anhydride
Vzorec: C₉H₆O₃
MW: 162.14
Storage Temperature: Ambient
MDL Number: MFCD00047316
CAS číslo: 4792-30-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR