Tisk…

4-(p-Tolyl)morpholine 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Morpholinotoluene

OR925167-1G OR925167-5G
APOSOR925167-1GEA 1700 CZK
APOSOR925167-1G APOSOR925167-5G
4-(p-Tolyl)morpholine 95%
4-(p-Tolyl)morpholine
Vzorec: C₁₁H₁₅NO
MW: 177.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00222003
CAS číslo: 3077-16-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR