Tisk…

2-Benzylimidazoline 95%

Dodavatel: Apollo Scientific

OR925139-25G OR925139-5G
APOSOR925139-25GEA 14290 CZK
APOSOR925139-25G APOSOR925139-5G
2-Benzylimidazoline 95%
2-Benzylimidazoline
Vzorec: C₁₀H₁₂N₂
Storage Temperature: Ambient
MDL Number: MFCD00005182
CAS číslo: 59-98-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR