Tisk…

3,3-Dimethyl-1,2-epoxybutane 95%

Dodavatel: Apollo Scientific
Nebezpečí

OR925110-1G OR925110-5G
APOSOR925110-1GEA 2900 CZK
APOSOR925110-1G APOSOR925110-5G
3,3-Dimethyl-1,2-epoxybutane 95%
3,3-Dimethyl-1,2-epoxybutane
Vzorec: C₆H₁₂O
MW: 100,16 g/mol
Bod varu: 95…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00051590
CAS číslo: 2245-30-9
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR