Tisk…

2,4-Dimethylphenyl isocyanate 95%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,4-Xylyl isocyanate, 1-Isocyanato-2,4-dimethylbenzene

OR925058-1G OR925058-25G OR925058-5G
APOSOR925058-1GEA 1080 CZK
APOSOR925058-1G APOSOR925058-25G APOSOR925058-5G
2,4-Dimethylphenyl isocyanate 95%
2,4-Dimethylphenyl isocyanate

  • May form small amount of sediment.

Vzorec: C₉H₉NO
MW: 147,18 g/mol
Bod varu: 114…155 °C
Hustota: 1,041 g/cm³ (25 °C)
Bod vzplanutí: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00013853
CAS číslo: 51163-29-2
UN: 2206
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR