Tisk…

2-Methyl-4-phenyl-2-butanol 99%

Dodavatel: Apollo Scientific

Synonyms: 2-Phenylethyldimethylcarbinol, Dimethyl-beta-phenylethylcarbinol

OR925050-500G
APOSOR925050-500GEA 3870 CZK
APOSOR925050-500G
2-Methyl-4-phenyl-2-butanol 99%
2-Methyl-4-phenyl-2-butanol
Vzorec: C₁₁H₁₆O
MW: 164,25 g/mol
Bod tání: 30 °C
Storage Temperature: Ambient
MDL Number: MFCD00004472
CAS číslo: 103-05-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR