(3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol 95%
Dodavatel: Apollo Scientific
OR925042-1G
OR925042-5G
APOSOR925042-1GEA
8070
CZK
APOSOR925042-1G
APOSOR925042-5G
(3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol 95%
(3,4-Dihydro-2H-pyran-6-yl)dimethylsilanol
Vzorec:
C₇H₁₄O₂Si MW: 158,27 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09265116 CAS číslo: 304669-35-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...