Tisk…

Ethyl-6-oxaspiro[2.5]octane-1-carboxylate

Dodavatel: Apollo Scientific

OR924935-10G OR924935-1G OR924935-5G
APOSOR924935-10GEA 41000 CZK
APOSOR924935-10G APOSOR924935-1G APOSOR924935-5G
Ethyl-6-oxaspiro[2.5]octane-1-carboxylate
Ethyl-6-oxaspiro[2.5]octane-1-carboxylate
Vzorec: C₁₀H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD23105848
CAS číslo: 909406-74-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR