Tisk…

N-(2-Aminoethyl)-N-ethyl-m-toluidine 95%

Dodavatel: Apollo Scientific

OR924816-5G
APOSOR924816-5GEA 6580 CZK
APOSOR924816-5G
N-(2-Aminoethyl)-N-ethyl-m-toluidine 95%
N-(2-Aminoethyl)-N-ethyl-m-toluidine
Vzorec: C₁₁H₁₈N₂
MW: 178,28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00025603
CAS číslo: 19248-13-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR