Tisk…

2,2-Dimethyl-1,3-propanediol 95%

Dodavatel: Apollo Scientific

Synonyms: Neopentyl Glycol, 2,2-Dimethylpropane-1,3-diol

OR924723-500G
APOSOR924723-500GEA 0 CZK
APOSOR924723-500G
2,2-Dimethyl-1,3-propanediol 95%
2,2-Dimethyl-1,3-propanediol
Vzorec: C₅H₁₂O₂
MW: 104,15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004685
CAS číslo: 126-30-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR