Tisk…

Mucochloric acid 99%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 2,3-Dichloromalealdehydic acid

OR924576-500G
APOSOR924576-500GEA 3580 CZK
APOSOR924576-500G
Mucochloric acid 99%
Mucochloric acid
Vzorec: C₄H₂Cl₂O₃
MW: 168,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135465
CAS číslo: 87-56-9
UN: 2923
ADR: 8+6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR