Tisk…

3-Propoxypropylamine 95%

Dodavatel: Apollo Scientific

Synonyms: 1-Amino-3-propoxypropane

OR924493-1G OR924493-5G
APOSOR924493-1GEA 2610 CZK
APOSOR924493-1G APOSOR924493-5G
3-Propoxypropylamine 95%
3-Propoxypropylamine
Vzorec: C₆H₁₅NO
MW: 117,19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00129001
CAS číslo: 16728-59-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR