Tisk…

4-(Methylthio)benzonitrile 95%

Dodavatel: Apollo Scientific

OR924462-25G OR924462-5G
APOSOR924462-25GEA 4800 CZK
APOSOR924462-25G APOSOR924462-5G
4-(Methylthio)benzonitrile 95%
4-(Methylthio)benzonitrile
Vzorec: C₈H₇NS
MW: 149,22 g/mol
Bod varu: 272 °C
Bod tání: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00010381
CAS číslo: 21382-98-9
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR