Tisk…

1-Phenyl-1-butanol 95%

Dodavatel: Apollo Scientific

OR924460-25G OR924460-5G
APOSOR924460-25GEA 9100 CZK
APOSOR924460-25G APOSOR924460-5G
1-Phenyl-1-butanol 95%
1-Phenyl-1-butanol
Vzorec: C₁₀H₁₄O
MW: 150,2176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066173
CAS číslo: 614-14-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR