Tisk…

trans-4-Methoxycinnamaldehyde 95%

Dodavatel: Apollo Scientific

OR924215-100G OR924215-25G
APOSOR924215-100GEA 6090 CZK
APOSOR924215-100G APOSOR924215-25G
trans-4-Methoxycinnamaldehyde 95%
trans-4-Methoxycinnamaldehyde
Vzorec: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00017343
CAS číslo: 24680-50-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR