Tisk…

N-Ethylpropionamide 95%

Dodavatel: Apollo Scientific

OR924195-25G OR924195-5G
APOSOR924195-25GEA 9530 CZK
APOSOR924195-25G APOSOR924195-5G
N-Ethylpropionamide 95%
N-Ethylpropionamide
Vzorec: C₅H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD00059388
CAS číslo: 5129-72-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR