Tisk…

Vinyltrimethoxysilane 97%

Dodavatel: Apollo Scientific

OR924149-100G OR924149-1KG OR924149-500G
APOSOR924149-100GEA 3540 CZK
APOSOR924149-100G APOSOR924149-1KG APOSOR924149-500G
Vinyltrimethoxysilane 97%
Vinyltrimethoxysilane
Vzorec: C₅H₁₂O₃Si
MW: 148,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00008605
CAS číslo: 2768-02-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR