Tisk…

Ethyl isoxazole-3-carboxylate 95%

Dodavatel: Apollo Scientific

OR924041-1G OR924041-5G
APOSOR924041-1GEA 0 CZK
APOSOR924041-1G APOSOR924041-5G
Ethyl isoxazole-3-carboxylate 95%
Ethyl isoxazole-3-carboxylate
Vzorec: C₆H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD08234628
CAS číslo: 3209-70-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR