Tisk…

Allyl ethyl carbonate 95%

Dodavatel: Apollo Scientific

Synonyms: Allyl Ethyl Carbonate, Carbonicacidallylethylester, Carbonic acid allyl ethyl ester, Carbonic acid Allyl Ethyl Ester

OR923998-10G OR923998-5G
APOSOR923998-10GEA 0 CZK
APOSOR923998-10G APOSOR923998-5G
Allyl ethyl carbonate 95%
Allyl ethyl carbonate
Vzorec: C₆H₁₀O₃
MW: 130,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00049113
CAS číslo: 1469-70-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR