Tisk…

4-Nitro-1H-indole-3-carbaldehyde 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Nitroindole-3-carboxaldehyde

OR923916-1G OR923916-5G
APOSOR923916-1GEA 4270 CZK
APOSOR923916-1G APOSOR923916-5G
4-Nitro-1H-indole-3-carbaldehyde 95%
4-Nitro-1H-indole-3-carbaldehyde
Vzorec: C₉H₆N₂O₃
MW: 190.158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04973979
CAS číslo: 10553-11-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR