Tisk…

4,5,6,7-Tetrahydro-2H-indazol-3-ol 96%

Dodavatel: Apollo Scientific

OR923882-1G OR923882-5G
APOSOR923882-1GEA 5390 CZK
APOSOR923882-1G APOSOR923882-5G
4,5,6,7-Tetrahydro-2H-indazol-3-ol 96%
4,5,6,7-Tetrahydro-2H-indazol-3-ol
Vzorec: C₇H₁₀N₂O
Storage Temperature: Ambient
MDL Number: MFCD00462221
CAS číslo: 402602-24-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR