Tisk…

2'-Methyl-3'-nitroacetanilide 95%

Dodavatel: Apollo Scientific

Synonyms: N-(2-Methyl-3-nitrophenyl)acetamide

OR923868-25G OR923868-5G
APOSOR923868-25GEA 6520 CZK
APOSOR923868-25G APOSOR923868-5G
2'-Methyl-3'-nitroacetanilide 95%
2'-Methyl-3'-nitroacetanilide
Vzorec: C₉H₁₀N₂O₃
MW: 194,190 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01047363
CAS číslo: 56207-36-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR