Tisk…

2,5-Dichlorobenzyl alcohol 95%

Dodavatel: Apollo Scientific

Synonyms: (2,5-Dichlorophenyl)methanol

OR923844-25G
APOSOR923844-25GEA 2650 CZK
APOSOR923844-25G
2,5-Dichlorobenzyl alcohol 95%
2,5-Dichlorobenzyl alcohol
Vzorec: C₇H₆Cl₂O
MW: 177,03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004607
CAS číslo: 34145-05-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR