Tisk…

2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester 95%

Dodavatel: Apollo Scientific

Synonyms: N-Boc-2,5-diaza-bicyclo[2.2.1]heptane, tert-Butyl 2,5-diazabicyclo[2.2.1]heptane-2-carboxylate

OR923781-1G OR923781-5G
APOSOR923781-1GEA 1620 CZK
APOSOR923781-1G APOSOR923781-5G
2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester 95%
2,5-Diazabicyclo[2.2.1]heptane-2-carboxylic acid tert-butyl ester
Vzorec: C₁₀H₁₈N₂O₂
MW: 198,27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06411671
CAS číslo: 198989-07-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR