Tisk…

(2-Methyl-2-propenyl)succinic anhydride 95%

Dodavatel: Apollo Scientific

Synonyms: 4-Methyl-4-pentene-1,2-dicarboxylic Anhydride, (beta-Methallyl)succinic Anhydride

OR923717-1G OR923717-25G OR923717-5G
APOSOR923717-1GEA 4140 CZK
APOSOR923717-1G APOSOR923717-25G APOSOR923717-5G
(2-Methyl-2-propenyl)succinic anhydride 95%
(2-Methyl-2-propenyl)succinic anhydride
Vzorec: C₈H₁₀O₃
MW: 154,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075477
CAS číslo: 18908-20-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR