Isobutyric anhydride 95%
Dodavatel: Apollo Scientific
OR923617-100G
OR923617-1KG
OR923617-500G
APOSOR923617-100GEA
3540
CZK
APOSOR923617-100G
APOSOR923617-1KG
APOSOR923617-500G
Isobutyric anhydride 95%
Isobutyric anhydride
Vzorec:
C₈H₁₄O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00008913 CAS číslo: 97-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...