Tisk…

1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde 95%

Dodavatel: Apollo Scientific

OR923536-1G OR923536-5G
APOSOR923536-1GEA 5200 CZK
APOSOR923536-1G APOSOR923536-5G
1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde 95%
1-Isopropyl-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Vzorec: C₉H₁₄N₂O
Storage Temperature: Ambient
MDL Number: MFCD06260699
CAS číslo: 890626-07-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR