2-(Ethylamino)pyridine 95%
Dodavatel: Apollo Scientific
OR923489-10G
OR923489-1G
OR923489-5G
APOSOR923489-10GEA
16340
CZK
APOSOR923489-10G
APOSOR923489-1G
APOSOR923489-5G
2-(Ethylamino)pyridine 95%
2-(Ethylamino)pyridine
Vzorec:
C₇H₁₀N₂ Storage Temperature: Ambient |
MDL Number:
MFCD04038418 CAS číslo: 37059-57-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...