Tisk…

2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 95%

Dodavatel: Apollo Scientific

OR923428-1G OR923428-5G
APOSOR923428-1GEA 5790 CZK
APOSOR923428-1G APOSOR923428-5G
2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 95%
2-(2-Methyl-1H-imidazol-1-yl)benzaldehyde
Vzorec: C₁₁H₁₀N₂O
MW: 186.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05864668
CAS číslo: 914348-86-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR