Tisk…

3-Chloropropionic acid 98%

Dodavatel: Apollo Scientific

Synonyms: 3-Chloropropanoic acid

OR923420-100G OR923420-1KG OR923420-500G
APOSOR923420-100GEA 1490 CZK
APOSOR923420-100G APOSOR923420-1KG APOSOR923420-500G
3-Chloropropionic acid 98%
3-Chloropropionic acid
Vzorec: C₃H₅ClO₂
MW: 108,52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002764
CAS číslo: 107-94-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR