Tisk…

Ethyl glycidyl ether 95%

Dodavatel: Apollo Scientific

OR923221-100G OR923221-25G
APOSOR923221-100GEA 12400 CZK
APOSOR923221-100G APOSOR923221-25G
Ethyl glycidyl ether 95%
Ethyl glycidyl ether
Vzorec: C₅H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00085264
CAS číslo: 4016-11-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR