Tisk…

4-Phenoxybenzoyl chloride 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR9232-1G OR9232-5G
APOSOR9232-1GEA 6620 CZK
APOSOR9232-1G APOSOR9232-5G
4-Phenoxybenzoyl chloride 97%
4-Phenoxybenzoyl chloride
Vzorec: C₁₃H₉ClO₂
Bod tání: 22…23 °C
Storage Temperature: Ambient
MDL Number: MFCD00043966
CAS číslo: 1623-95-6
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR