3-Phenoxytoluene 95%
Dodavatel: Apollo Scientific
Synonyms:
Phenyl m-tolyl ether, 1-Methyl-3-phenoxybenzene
OR923189-100G
OR923189-500G
APOSOR923189-100GEA
2570
CZK
APOSOR923189-100G
APOSOR923189-500G
3-Phenoxytoluene 95%
3-Phenoxytoluene
Vzorec:
C₁₃H₁₂O MW: 184,24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00008531 CAS číslo: 3586-14-9 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...