Tisk…

S-Butyl thiobenzoate 95%

Dodavatel: Apollo Scientific

OR923124-1G OR923124-25G OR923124-5G
APOSOR923124-1GEA 4370 CZK
APOSOR923124-1G APOSOR923124-25G APOSOR923124-5G
S-Butyl thiobenzoate 95%
S-Butyl thiobenzoate
Vzorec: C₁₁H₁₄OS
Storage Temperature: Ambient
MDL Number: MFCD00039955
CAS číslo: 7269-35-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR