Benzo[1,3]dioxol-5-ylacetyl chloride 95%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR9231-1G
OR9231-250MG
APOSOR9231-1GEA
3110
CZK
APOSOR9231-1G
APOSOR9231-250MG
Benzo[1,3]dioxol-5-ylacetyl chloride 95%
Benzo[1,3]dioxol-5-ylacetyl chloride
Vzorec:
C₉H₇ClO₃ MW: 198.61 g/mol Bod varu: 110…112 °C/0.75mmHg Storage Temperature: Ambient |
MDL Number:
MFCD03424722 CAS číslo: 6845-81-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...