Tisk…

Ethyl 4-hydroxybenzoate 96%

Dodavatel: Apollo Scientific

Synonyms: Ethyl-4-hydroxybenzoat, Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethyl-p-hydroxybenzoat, Ethylparaben

OR923091-500G
APOSOR923091-500GEA 744 CZK
APOSOR923091-500G
Ethyl 4-hydroxybenzoate 96%
Ethyl 4-hydroxybenzoate
Vzorec: C₉H₁₀O₃
MW: 166,18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002353
CAS číslo: 120-47-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR