Tisk…

N-Butylpropionamide 95%

Dodavatel: Apollo Scientific

OR923016-25G OR923016-5G
APOSOR923016-25GEA 13230 CZK
APOSOR923016-25G APOSOR923016-5G
N-Butylpropionamide 95%
N-Butylpropionamide
Vzorec: C₇H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD00059416
CAS číslo: 2955-67-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR