Tisk…

1-Piperidin-4-ylethanoltrifluoroacetate 95%

Dodavatel: Apollo Scientific

Synonyms: 1-(Piperidin-4-yl)ethan-1-ol

OR922934-1G OR922934-250MG
APOSOR922934-1GEA 9920 CZK
APOSOR922934-1G APOSOR922934-250MG
1-Piperidin-4-ylethanoltrifluoroacetate 95%
1-Piperidin-4-ylethanoltrifluoroacetate
Vzorec: C₉H₁₆F₃NO₃
MW: 243,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17676096
CAS číslo: 6457-48-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR