Tisk…

3-Isopropoxyaniline 95%

Dodavatel: Apollo Scientific

OR922907-5G
APOSOR922907-5GEA 1210 CZK
APOSOR922907-5G
3-Isopropoxyaniline 95%
3-Isopropoxyaniline
Vzorec: C₉H₁₃NO
MW: 151,21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00052700
CAS číslo: 41406-00-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR