Butyl nicotinate 98%
Dodavatel: Apollo Scientific
Synonyms:
Nicotinic acid butyl ester, Butyl 3-pyridinecarboxylate, n-Butyl nicotinate
OR922897-100G
APOSOR922897-100GEA
1040
CZK
APOSOR922897-100G
Butyl nicotinate 98%
Butyl nicotinate
Vzorec:
C₁₀H₁₃NO₂ MW: 179,22 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00006390 CAS číslo: 6938-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...