Tisk…

Chloroacetaldehyde dimethyl acetal 96%

Dodavatel: Apollo Scientific

OR922884-100G OR922884-1KG OR922884-500G
APOSOR922884-100GEA 1910 CZK
APOSOR922884-100G APOSOR922884-1KG APOSOR922884-500G
Chloroacetaldehyde dimethyl acetal 96%
Chloroacetaldehyde dimethyl acetal
Vzorec: C₄H₉ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00000948
CAS číslo: 97-97-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR