Tisk…
Fumaric acid monoisopropyl ester 95%
Dodavatel: Apollo Scientific
Synonyms: Monoisopropyl fumarate
OR922855-25G OR922855-5G OR922855-100G
APOSOR922855-25GEA 6020 CZK
APOSOR922855-25G APOSOR922855-5G APOSOR922855-100G
Fumaric acid monoisopropyl ester 95%
Fumaric acid monoisopropyl ester
Vzorec: C₇H₁₀O₄
MW: 158,15 g/mol
Storage Temperature: Ambient
CAS číslo: 7529-87-5