Tisk…

Methyl sorbate 98%

Dodavatel: Apollo Scientific

OR922849-100G OR922849-25G
APOSOR922849-100GEA 10550 CZK
APOSOR922849-100G APOSOR922849-25G
Methyl sorbate 98%
Methyl sorbate
Vzorec: C₇H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00038319
CAS číslo: 1515-80-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR